siRNA Details
| Gene Symbol: Ccr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009915.2 | NP_034045.1 | 3 | 165 | ||
| GenBank mRNA | AK046579.1 | - | 399 | |||
| AK138599.1 | - | 2 | 478 | |||
| BC138803.1 | - | 2 | 125 | 8861825 | ||
| BC138804.1 | - | 2 | 125 | 8861826 | ||
| U47035.1 | - | 2 | 74 | |||
| U51717.1 | - | 2 | 123 | |||
| U56819.1 | - | 3 | 165 | |||

Ccr2
Genomic Map