siRNA Details
| Gene Symbol: Cnih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009919.2 | NP_034049.2 | 2 | 198 | ||
| GenBank mRNA | AF022811.1 | - | 2 | 189 | ||
| AK002293.1 | - | 2 | 166 | |||
| AK011200.1 | - | 2 | 151 | |||
| AK152432.1 | - | 2 | 149 | |||
| AK154479.1 | - | 2 | 149 | |||
| AK168154.1 | - | 2 | 149 | |||
| BC034868.1 | - | 2 | 158 | 2938052 | ||
| DV653213.1 | - | 2 | 252 | |||

Cnih1
Genomic Map