siRNA Details
| Gene Symbol: Hps3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146323.1 | NP_001139795.1 | 12 | 1791 | ||
| NM_080634.4 | NP_542365.3 | 12 | 2187 | |||
| GenBank mRNA | AF393747.1 | - | 12 | 2176 | ||
| AF393780.1 | - | 12 | 2179 | |||
| AK034377.1 | - | 12 | 2158 | |||
| AK036663.1 | - | 12 | 2195 | |||
| AK155497.1 | - | 12 | 2154 | |||
| AK157953.1 | - | 12 | 2173 | |||
| AK169855.1 | - | 11 | 2259 | |||
| AK170581.1 | - | 12 | 1703 | |||
| AK171997.1 | - | 12 | 1779 | |||
| BC020105.1 | - | 2 | 286 | |||
| BC038087.1 | - | 2 | 278 | |||
| BC049350.1 | - | 3 | 378 | |||
| BC058235.1 | - | 12 | 2161 | 6307400 | ||

Hps3
Genomic Map