siRNA Details
| Gene Symbol: Cobl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282993.1 | NP_001269922.1 | 11 | 3369 | ||
| NM_001282994.1 | NP_001269923.1 | 12 | 3275 | |||
| NM_172496.3 | NP_766084.3 | 12 | 3444 | |||
| GenBank mRNA | AK122334.1 | - | 12 | 3275 | ||
| AK147366.1 | - | 11 | 3369 | |||
| AK147633.1 | - | 11 | 3369 | |||
| AY308745.1 | - | 12 | 3284 | |||
| BC023264.1 | - | 1 | 446 | |||
| BC060061.1 | - | 1 | 1008 | |||
| BC145116.1 | - | 9 | 2749 | |||
| BC145118.1 | - | 12 | 3284 | 9053666 | ||
| BC150706.1 | - | 12 | 3284 | 9087617 | ||

Cobl
Genomic Map