siRNA Details
| Gene Symbol: Col11a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009926.1 | NP_034056.1 | 63 | 4999 | ||
| GenBank mRNA | KJ650220.1 | - | 64 | 4976 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col11a2| Gene Symbol: Col11a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009926.1 | NP_034056.1 | 63 | 4999 | ||
| GenBank mRNA | KJ650220.1 | - | 64 | 4976 | ||