siRNA Details
Gene Symbol: Col11a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009926.1 | NP_034056.1 | 19 | 1791 | ||
GenBank mRNA | AK138017.1 | - | 14 | 1059 | ||
D38412.1 | - | 21 | 2013 | |||
KJ650220.1 | - | 20 | 1768 | |||
U16789.1 | - | 19 | 1792 |
