siRNA Details
| Gene Symbol: Col4a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163155.1 | NP_001156627.1 | 52, 53 | 5312 | ||
| NM_007736.4 | NP_031762.2 | 53, 54 | 5140 | |||
| GenBank mRNA | AB041350.1 | - | 52, 53 | 5170 | ||
| AK080682.1 | - | 20, 21 | 2321 | |||
| AK133179.1 | - | 53, 54 | 5141 | |||
| AK164687.1 | - | 12, 13 | 1384 | |||
| BC016479.1 | - | 28, 29 | 3077 | |||
| BC043317.1 | - | 19, 20 | 1958 | |||
| BC082788.1 | - | 52, 53 | 5151 | 5343488 | ||
| Z35168.1 | - | 4, 5 | 682 | |||

Col4a5
Genomic Map