siRNA Details
Gene Symbol: Col4a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163155.1 | NP_001156627.1 | 22 | 1761 | ||
NM_007736.4 | NP_031762.2 | 23 | 1589 | |||
GenBank mRNA | AB041350.1 | - | 22 | 1619 | ||
AK133179.1 | - | 23 | 1590 | |||
BC082788.1 | - | 22 | 1600 | 5343488 |
