siRNA Details
| Gene Symbol: Col6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146007.2 | NP_666119.1 | 21 | 1837 | ||
| GenBank mRNA | AK044870.1 | - | 21 | 1759 | ||
| AK128938.1 | - | 21 | 1760 | |||
| AK132540.1 | - | 21 | 1761 | |||
| BC034414.1 | - | 21 | 1747 | 3594974 | ||
| L06343.1 | - | 17 | 860 | |||
| X62332.1 | - | 17 | 860 | |||
| X65582.1 | - | 20 | 1676 | |||

Col6a2
Genomic Map