siRNA Details
Gene Symbol: Col6a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146007.2 | NP_666119.1 | 3, 4 | 926 | ||
GenBank mRNA | AK042826.1 | - | 3, 4 | 848 | ||
AK044870.1 | - | 3, 4 | 848 | |||
AK128938.1 | - | 3, 4 | 849 | |||
AK132540.1 | - | 3, 4 | 847 | |||
BC034414.1 | - | 3, 4 | 836 | 3594974 | ||
X65582.1 | - | 2, 3 | 765 |
