siRNA Details
| Gene Symbol: Coq7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF053770.1 | - | 1 | 106 | ||
| AF080580.1 | - | 2 | 169 | |||
| AF098949.1 | - | 2 | 235 | |||
| AK137841.1 | - | 2 | 233 | |||
| AK158380.1 | - | 2 | 220 | |||
| BC038681.1 | - | 2 | 202 | 1380996 | ||
| BY284216.1 | - | 2 | 274 | |||

Coq7
Genomic Map