siRNA Details
| Gene Symbol: Cp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276248.1 | NP_001263177.1 | 14 | 2748 | ||
| NM_001276250.1 | NP_001263179.1 | 14 | 2748 | |||
| NM_007752.3 | NP_031778.2 | 14 | 2751 | |||
| GenBank mRNA | AK014835.1 | - | 14 | 2734 | ||
| AK043248.1 | - | 14 | 2693 | |||
| AK080701.1 | - | 6 | 847 | |||
| AK160035.1 | - | 10 | 1677 | |||
| BC062957.1 | - | 14 | 2751 | 30547760 | ||
| DQ336396.1 | - | 14 | 2749 | |||
| U49430.1 | - | 14 | 2629 | |||

Cp
Genomic Map