siRNA Details
| Gene Symbol: Cpne6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136057.2 | NP_001129529.1 | 16 | 1886 | ||
| NM_001146183.1 | NP_001139655.1 | 16 | 1860 | |||
| NM_009947.3 | NP_034077.1 | 16 | 1689 | |||
| GenBank mRNA | AB008893.1 | - | 16 | 1689 | ||
| AK134548.1 | - | 16 | 1705 | |||
| AK158638.1 | - | 12 | 3687 | |||
| BC050766.1 | - | 16 | 1711 | 6591063 | ||
| BC051994.1 | - | 14 | 2343 | |||
| CO039302.1 | - | 2 | 331 | |||

Cpne6
Genomic Map