siRNA Details
| Gene Symbol: Cpne6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136057.2 | NP_001129529.1 | 7, 8 | 965 | ||
| NM_001146183.1 | NP_001139655.1 | 7, 8 | 942 | |||
| NM_009947.3 | NP_034077.1 | 7, 8 | 768 | |||
| GenBank mRNA | AB008893.1 | - | 7, 8 | 768 | ||
| AK134548.1 | - | 7, 8 | 787 | |||
| AK144410.1 | - | 7, 8 | 794 | |||
| AK158638.1 | - | 5, 6 | 2358 | |||
| BC050766.1 | - | 7, 8 | 790 | 6591063 | ||
| BC051994.1 | - | 6, 7 | 1350 | |||

Cpne6
Genomic Map