siRNA Details
| Gene Symbol: Cpox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007757.2 | NP_031783.2 | 3 | 960 | ||
| GenBank mRNA | AK157277.1 | - | 3 | 843 | ||
| AK166512.1 | - | 3 | 823 | |||
| BC017680.1 | - | 3 | 817 | 4241687 | ||
| CA535096.1 | - | 3 | 288 | |||
| D16333.1 | - | 3 | 874 | |||

Cpox
Genomic Map