siRNA Details
| Gene Symbol: Cpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009949.2 | NP_034079.2 | 5 | 1792 | ||
| GenBank mRNA | AK144441.1 | - | 5 | 1792 | ||
| AK150163.1 | - | 5 | 1764 | |||
| AK169038.1 | - | 5 | 1689 | |||
| BC138514.1 | - | 5 | 1727 | 8861534 | ||
| BC145859.1 | - | 5 | 1727 | 40131162 | ||
| U01163.1 | - | 5 | 1737 | |||

Cpt2
Genomic Map