siRNA Details
| Gene Symbol: Cr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007758.2 | NP_031784.1 | 18 | 3141 | ||
| GenBank mRNA | AK003072.2 | - | 18 | 3141 | ||
| AK007487.1 | - | 4 | 295 | |||
| BC109344.1 | - | 18 | 3074 | 40054528 | ||
| M29281.1 | - | 13 | 2145 | |||
| M35684.1 | - | 17 | 3015 | |||
| M61132.1 | - | 17 | 2982 | |||
| M81083.1 | - | 1 | 45 | |||

Cr2
Genomic Map