siRNA Details
| Gene Symbol: Pcdha6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007767.3 | NP_031793.2 | 1 | 2346 | ||
| GenBank mRNA | AY013764.1 | - | 1 | 2346 | ||
| BC054813.1 | - | 2 | 2355 | |||
| BC150815.1 | - | 1 | 2454 | 9087726 | ||
| BC150816.1 | - | 1 | 2454 | 9087727 | ||
| D86917.1 | - | 1 | 2472 | |||
| Gene Symbol: Gm38666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301256.1 | NP_001288185.1 | 1 | 2471 | ||

Pcdha6
Genomic Map