siRNA Details
| Gene Symbol: Crx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113330.1 | NP_001106801.1 | 4 | 970 | ||
| NM_007770.4 | NP_031796.1 | 4 | 998 | |||
| GenBank mRNA | AK053533.1 | - | 4 | 940 | ||
| AK139691.1 | - | 4 | 973 | |||
| BC016502.1 | - | 4 | 912 | 4527863 | ||
| U77615.1 | - | 4 | 951 | |||

Crx
Genomic Map