siRNA Details
| Gene Symbol: Cryba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009965.2 | NP_034095.1 | 4 | 318 | ||
| GenBank mRNA | AJ239052.1 | - | 4 | 318 | ||
| AK084145.1 | - | 1 | 895 | |||
| BC107342.2 | - | 4 | 318 | 40055280 | ||
| J00378.1 | - | 1 | 91 | |||
| V00728.1 | - | 2 | 210 | |||

Cryba1
Genomic Map