siRNA Details
Gene Symbol: Cryz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009968.3 | NP_034098.1 | 6 | 1172 | ||
GenBank mRNA | AK098082.1 | - | 6 | 656 | ||
AK133820.1 | - | 6 | 1173 | |||
AK134281.1 | - | 6 | 657 | |||
AK188548.1 | - | 3 | 179 | |||
BC003800.1 | - | 6 | 664 | 3492859 | ||
BC043076.1 | - | 6 | 633 | 6404996 | ||
D78646.1 | - | 3 | 199 | |||
S70056.1 | - | 6 | 684 |
