siRNA Details
| Gene Symbol: Csf3r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252651.1 | NP_001239580.1 | 4, 5 | 608 | ||
| NM_007782.3 | NP_031808.2 | 4, 5 | 608 | |||
| GenBank mRNA | AK089305.1 | - | 4, 5 | 553 | ||
| AK144318.1 | - | 3, 4 | 509 | |||
| BC115865.1 | - | 3, 4 | 485 | 40081337 | ||
| BC116635.1 | - | 3, 4 | 485 | 40081333 | ||
| M58288.1 | - | 4, 5 | 531 | |||

Csf3r
Genomic Map