siRNA Details
Gene Symbol: Ctse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007799.3 | NP_031825.2 | 9, 10 | 1384 | ||
GenBank mRNA | AK143581.1 | - | 8, 9 | 1086 | ||
AK145875.1 | - | 9, 10 | 1241 | |||
AK157907.1 | - | 8, 9 | 1106 | |||
AK165271.1 | - | 8, 9 | 1052 | |||
BC005432.1 | - | 9, 10 | 1384 | 3601519 | ||
CT010252.1 | - | 8, 9 | 1020 | |||
X97399.1 | - | 8, 9 | 1071 |
