siRNA Details
Gene Symbol: Ctsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007802.4 | NP_031828.2 | 6 | 763 | ||
GenBank mRNA | AK003425.1 | - | 6 | 688 | ||
AK132648.1 | - | 6 | 693 | |||
BC046320.1 | - | 6 | 717 | 6390854 | ||
X94444.1 | - | 5 | 635 |
