siRNA Details
| Gene Symbol: Cybb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007807.5 | NP_031833.3 | 6, 7 | 741 | ||
| GenBank mRNA | AK033570.1 | - | 6, 7 | 685 | ||
| AK053920.1 | - | 6, 7 | 685 | |||
| AK152257.1 | - | 6, 7 | 712 | |||
| AK153154.1 | - | 6, 7 | 712 | |||
| AK153259.1 | - | 6, 7 | 712 | |||
| AK154126.1 | - | 6, 7 | 741 | |||
| AK160031.1 | - | 1, 2 | 143 | |||
| AK171526.1 | - | 6, 7 | 727 | |||
| BC042838.1 | - | 6, 7 | 708 | |||
| BC071229.1 | - | 6, 7 | 696 | 4979804 | ||
| FJ168469.1 | - | 6, 7 | 730 | |||
| U43384.1 | - | 6, 7 | 667 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_023965, AJ295950, FQ217794, FQ231233.

Cybb
Genomic Map