siRNA Details
| Gene Symbol: Cyp19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007810.3 | NP_031836.1 | 7, 8 | 1063 | ||
| GenBank mRNA | AK143323.1 | - | 7, 8 | 1143 | ||
| BC103670.1 | - | 7, 8 | 1046 | 40047093 | ||
| BC104489.1 | - | 7, 8 | 1046 | 40047095 | ||
| D00659.1 | - | 7, 8 | 1068 | |||

Cyp19a1
Genomic Map