siRNA Details
| Gene Symbol: Cyp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136059.2 | NP_001129531.1 | 7 | 1538 | ||
| NM_009992.4 | NP_034122.1 | 7 | 1617 | |||
| GenBank mRNA | AK005000.1 | - | 7 | 1618 | ||
| BC125440.1 | - | 7 | 1537 | 40129955 | ||
| BC125444.1 | - | 7 | 1537 | 40129959 | ||
| K02588.1 | - | 7 | 1537 | |||
| Y00071.1 | - | 7 | 1538 | |||

Cyp1a1
Genomic Map