siRNA Details
| Gene Symbol: Cyp3a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007819.4 | NP_031845.1 | 11 | 1315 | ||
| GenBank mRNA | AK136494.1 | - | 11 | 1314 | ||
| AK136529.1 | - | 11 | 1316 | |||
| AK149572.1 | - | 11 | 1316 | |||
| BC046592.1 | - | 11 | 1294 | 6308234 | ||
| X63023.1 | - | 11 | 1291 | |||

Cyp3a13
Genomic Map