siRNA Details
| Gene Symbol: Cyp4b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007823.2 | NP_031849.1 | 12 | 1441 | ||
| GenBank mRNA | AK078882.1 | - | 10 | 1065 | ||
| AK143890.1 | - | 12 | 1454 | |||
| AK165445.1 | - | 12 | 1457 | |||
| BC008996.1 | - | 12 | 1445 | 3967680 | ||
| D50834.1 | - | 12 | 1441 | |||

Cyp4b1
Genomic Map