siRNA Details
| Gene Symbol: Dgka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016811.2 | NP_058091.2 | 23, 24 | 2445 | ||
| GenBank mRNA | AF085219.1 | - | 22, 23 | 2131 | ||
| AK008063.1 | - | 11, 12 | 1104 | |||
| AK008272.1 | - | 12, 13 | 1112 | |||
| AK008314.1 | - | 12, 13 | 1112 | |||
| BC006713.1 | - | 23, 24 | 2445 | 3584592 | ||
| JN832677.1 | - | 23, 24 | 2388 | |||
| JN832678.1 | - | 23, 24 | 2740 | |||
| JN832679.1 | - | 23, 24 | 2464 | |||

Dgka
Genomic Map