siRNA Details
| Gene Symbol: Daxx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199733.1 | NP_001186662.1 | 2 | 283 | ||
| NM_007829.4 | NP_031855.3 | 2 | 183 | |||
| GenBank mRNA | AF006040.1 | - | 1 | 37 | ||
| AK090356.1 | - | 2 | 114 | |||
| AK133863.1 | - | 2 | 91 | |||
| AK144332.1 | - | 2 | 114 | |||
| AK145906.1 | - | 2 | 114 | |||
| AK146743.1 | - | 2 | 110 | |||
| AK151279.1 | - | 2 | 103 | |||
| AK153149.1 | - | 2 | 71 | |||
| AK156425.1 | - | 2 | 284 | |||
| AK172616.1 | - | 2 | 284 | |||
| BC128373.1 | - | 2 | 96 | 40111681 | ||
| DV653043.1 | - | 2 | 130 | |||
| KJ650207.1 | - | 1 | 13 | |||

Daxx
Genomic Map