siRNA Details
| Gene Symbol: Dbnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146308.1 | NP_001139780.1 | 1 | 133 | ||
| NM_001146309.1 | NP_001139781.1 | 1 | 133 | |||
| NM_013810.3 | NP_038838.1 | 1 | 133 | |||
| GenBank mRNA | AK046073.1 | - | 1 | 113 | ||
| AK053796.1 | - | 1 | 129 | |||
| AK078082.1 | - | 1 | 100 | |||
| AK134136.1 | - | 1 | 100 | |||
| AK134487.1 | - | 1 | 100 | |||
| AK140752.1 | - | 1 | 100 | |||
| AK142818.1 | - | 1 | 83 | |||
| AK146920.1 | - | 1 | 100 | |||
| AK151096.1 | - | 1 | 113 | |||
| AK152110.1 | - | 1 | 100 | |||
| AK152204.1 | - | 1 | 113 | |||
| AK153072.1 | - | 1 | 113 | |||
| AK153389.1 | - | 1 | 87 | |||
| AK159984.1 | - | 1 | 110 | |||
| AK161973.1 | - | 1 | 113 | |||
| AK167956.1 | - | 1 | 73 | |||
| AK168898.1 | - | 1 | 110 | |||
| AK172471.1 | - | 1 | 90 | |||
| AY098595.1 | - | 1 | 56 | |||
| BC046430.1 | - | 1 | 98 | 4986658 | ||
| U58884.1 | - | 1 | 86 | |||

Dbnl
Genomic Map