siRNA Details
| Gene Symbol: Dbt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010022.3 | NP_034152.2 | 7 | 893 | ||
| GenBank mRNA | AK075831.1 | - | 7 | 875 | ||
| AK165959.1 | - | 7 | 903 | |||
| BC055890.1 | - | 7 | 849 | 4242586 | ||
| L42996.1 | - | 7 | 846 | |||

Dbt
Genomic Map