siRNA Details
| Gene Symbol: Dcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190451.1 | NP_001177380.1 | 2 | 357 | ||
| NM_007833.5 | NP_031859.1 | 2 | 265 | |||
| GenBank mRNA | AK031456.1 | - | 1 | 2691 | ||
| AK135287.1 | - | 2 | 249 | |||
| AK145905.1 | - | 2 | 247 | |||
| AK147020.1 | - | 2 | 249 | |||
| AK161788.1 | - | 2 | 247 | |||
| AK165408.1 | - | 2 | 247 | |||
| BC060126.1 | - | 1 | 78 | |||
| BC132521.1 | - | 2 | 116 | 40130798 | ||
| BC138564.1 | - | 2 | 116 | 8861584 | ||
| BG088078.2 | - | 2 | 357 | |||
| X53929.1 | - | 2 | 274 | |||

Dcn
Genomic Map