siRNA Details
| Gene Symbol: Dctn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198866.1 | NP_001185795.1 | 28 | 3324 | ||
| NM_001198867.1 | NP_001185796.1 | 24 | 3249 | |||
| NM_007835.2 | NP_031861.2 | 28 | 3570 | |||
| GenBank mRNA | AK157867.1 | - | 27 | 3457 | ||
| AK171376.1 | - | 24 | 3236 | |||
| AK172261.1 | - | 24 | 3223 | |||
| BC066061.1 | - | 28 | 3297 | 6849060 | ||
| U60312.1 | - | 28 | 3305 | |||

Dctn1
Genomic Map