siRNA Details
| Gene Symbol: Dctn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198866.1 | NP_001185795.1 | 22, 23 | 2656 | ||
| NM_001198867.1 | NP_001185796.1 | 19, 20 | 2596 | |||
| NM_007835.2 | NP_031861.2 | 22, 23 | 2902 | |||
| GenBank mRNA | AK157867.1 | - | 22, 23 | 2903 | ||
| AK171376.1 | - | 19, 20 | 2583 | |||
| AK172261.1 | - | 19, 20 | 2570 | |||
| AK199428.1 | - | 1, 2 | 115 | |||
| BC066061.1 | - | 22, 23 | 2629 | 6849060 | ||
| U60312.1 | - | 22, 23 | 2637 | |||

Dctn1
Genomic Map