siRNA Details
| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007838.2 | NP_031864.2 | 10 | 1237 | ||
| GenBank mRNA | AK077651.1 | - | 10 | 1099 | ||
| AK081542.1 | - | 10 | 1129 | |||
| AK084267.1 | - | 10 | 1121 | |||
| AK148095.1 | - | 10 | 1118 | |||
| AK150683.1 | - | 10 | 1128 | |||
| BC068132.1 | - | 10 | 1237 | 6808198 | ||
| D89063.1 | - | 10 | 1239 | |||

Ddost
Genomic Map