siRNA Details
| Gene Symbol: Ackr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010045.2 | NP_034175.2 | 2 | 523 | ||
| GenBank mRNA | AF016584.1 | - | 2 | 464 | ||
| AF109159.1 | - | 2 | 523 | |||
| AY289754.1 | - | 2 | 430 | |||
| AY291295.1 | - | 2 | 430 | |||
| BC005583.1 | - | 2 | 522 | 3968984 | ||

Ackr1
Genomic Map