siRNA Details
Gene Symbol: Slc25a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153150.2 | NP_694790.1 | 3, 4 | 445 | ||
GenBank mRNA | AK005070.1 | - | 3, 4 | 375 | ||
AK159289.1 | - | 3, 4 | 408 | |||
AK170191.1 | - | 3, 4 | 409 | |||
BC037087.1 | - | 3, 4 | 405 | 5124403 |
