siRNA Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK156391.1 | - | 11 | 1461 | ||
| AK220466.1 | - | 24 | 3210 | |||
| BC070412.1 | - | 25 | 3375 | 5705112 | ||
| U96963.1 | - | 23 | 3213 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Diap1| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK156391.1 | - | 11 | 1461 | ||
| AK220466.1 | - | 24 | 3210 | |||
| BC070412.1 | - | 25 | 3375 | 5705112 | ||
| U96963.1 | - | 23 | 3213 | |||