siRNA Details
| Gene Symbol: Dffb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007859.4 | NP_031885.3 | 6, 7 | 878 | ||
| GenBank mRNA | AB009377.1 | - | 6, 7 | 786 | ||
| AK077619.1 | - | 6, 7 | 924 | |||
| AK089267.1 | - | 6, 7 | 856 | |||
| BC053052.1 | - | 6, 7 | 808 | 6403143 | ||

Dffb
Genomic Map