siRNA Details
| Gene Symbol: Dio1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007860.3 | NP_031886.3 | 1 | 275 | ||
| GenBank mRNA | AK002549.1 | - | 1 | 276 | ||
| BC089608.1 | - | 1 | 275 | 6766076 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dio1| Gene Symbol: Dio1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007860.3 | NP_031886.3 | 1 | 275 | ||
| GenBank mRNA | AK002549.1 | - | 1 | 276 | ||
| BC089608.1 | - | 1 | 275 | 6766076 | ||