siRNA Details
| Gene Symbol: Dmwd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010058.2 | NP_034188.2 | 2 | 514 | ||
| GenBank mRNA | AK086899.1 | - | 1 | 74 | ||
| BC089027.1 | - | 2 | 746 | 30615314 | ||
| CF169673.1 | - | 2 | 177 | |||
| S60312.1 | - | 2 | 154 | |||

Dmwd
Genomic Map