siRNA Details
Gene Symbol: Dmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007868.5 | NP_031894.1 | 21 | 2972 | ||
GenBank mRNA | M18025.1 | - | 13 | 1830 | ||
M68859.1 | - | 21 | 2945 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007868.5 | NP_031894.1 | 21 | 2972 | ||
GenBank mRNA | M18025.1 | - | 13 | 1830 | ||
M68859.1 | - | 21 | 2945 |