siRNA Details
| Gene Symbol: Dnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301737.1 | NP_001288666.1 | 5 | 735 | ||
| NM_010065.3 | NP_034195.2 | 5 | 735 | |||
| GenBank mRNA | AK011651.1 | - | 5 | 702 | ||
| AK144142.1 | - | 5 | 686 | |||
| AK220483.1 | - | 5 | 677 | |||
| BC034679.1 | - | 5 | 689 | 4506865 | ||
| BC058623.1 | - | 5 | 669 | 5706803 | ||
| CA328097.1 | - | 3 | 361 | |||
| L31395.1 | - | 5 | 632 | |||
| L31397.1 | - | 5 | 688 | |||

Dnm1
Genomic Map