siRNA Details
| Gene Symbol: Doc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007873.2 | NP_031899.2 | 4 | 748 | ||
| GenBank mRNA | BC067030.1 | - | 4 | 748 | 30534150 | |
| D85037.1 | - | 4 | 740 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Doc2b| Gene Symbol: Doc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007873.2 | NP_031899.2 | 4 | 748 | ||
| GenBank mRNA | BC067030.1 | - | 4 | 748 | 30534150 | |
| D85037.1 | - | 4 | 740 | |||