siRNA Details
| Gene Symbol: Dok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291799.1 | NP_001278728.1 | 4, 5 | 679 | ||
| NM_010070.4 | NP_034200.4 | 4, 5 | 681 | |||
| GenBank mRNA | AA790529.1 | - | 2, 3 | 216 | ||
| AK136382.1 | - | 4, 5 | 682 | |||
| BC013066.1 | - | 4, 5 | 675 | 3498170 | ||
| U78818.2 | - | 4, 5 | 646 | |||

Dok1
Genomic Map