siRNA Details
| Gene Symbol: Dok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291799.1 | NP_001278728.1 | 4 | 606 | ||
| NM_010070.4 | NP_034200.4 | 4 | 608 | |||
| GenBank mRNA | AA790529.1 | - | 2 | 143 | ||
| AK136382.1 | - | 4 | 609 | |||
| AK169871.1 | - | 4 | 604 | |||
| BC013066.1 | - | 4 | 602 | 3498170 | ||
| CJ053718.1 | - | 4 | 609 | |||
| U78818.2 | - | 4 | 573 | |||

Dok1
Genomic Map