siRNA Details
| Gene Symbol: Dok1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291799.1 | NP_001278728.1 | 4 | 554 | ||
| NM_010070.4 | NP_034200.4 | 4 | 556 | |||
| GenBank mRNA | AA790529.1 | - | 2 | 91 | ||
| AK136382.1 | - | 4 | 557 | |||
| AK169871.1 | - | 4 | 552 | |||
| BC013066.1 | - | 4 | 550 | 3498170 | ||
| CJ053718.1 | - | 4 | 557 | |||
| U78818.2 | - | 4 | 521 | |||

Dok1
Genomic Map