siRNA Details
| Gene Symbol: Dpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159543.1 | NP_001153015.1 | 9 | 1132 | ||
| NM_010074.3 | NP_034204.1 | 9 | 1132 | |||
| GenBank mRNA | AK040409.1 | - | 8 | 838 | ||
| AK085370.1 | - | 9 | 715 | |||
| AK153939.1 | - | 9 | 746 | |||
| AK163085.1 | - | 9 | 1137 | |||
| AK164775.1 | - | 9 | 715 | |||
| BC022183.1 | - | 9 | 994 | 3982742 | ||
| X58384.1 | - | 9 | 809 | |||

Dpp4
Genomic Map